Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 013199.09
also known as 013199-09
Price (USD)/ Each
410.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
410.00
Add to cart
Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number013199.09
Price (USD)/ Each
410.00
-
Add to cart
Chemical Identifiers
CAS14589-33-4
IUPAC Nameholmium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21HoO6
InChI KeyUEKRGRZSLATUQV-UHFFFAOYSA-N
SMILES[Ho+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofAHolmium: ≥29.0%
Appearance (Color)Yellow
CommentWaters of hydration: Report
It is used for Building Blocks of chemicals and , Chemical Synthesis, used as a trihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only