Holmium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Holmium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 013203.18
also known as 013203-18
Price (USD)/ Each
207.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
207.00
Add to cart
Holmium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number013203.18
Price (USD)/ Each
207.00
-
Add to cart
Chemical Identifiers
CAS223911-98-6
IUPAC Nameholmium(3+) tribromide
Molecular FormulaBr3Ho
InChI KeyMZNSYJWLQLXLHE-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only