Thermo Scientific Chemicals

Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013204.14
25 g, Each
Thermo Scientific Chemicals

Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013204.14
25 g, Each
Quantity
Catalog number: 013204.14
also known as 013204-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64438-54-6
IUPAC Name
gadolinium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21GdO6
InChI Key
PJCXQIZIMGZZIT-UHFFFAOYSA-N
SMILES
[Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

It is used as a laboratory chemicals & for manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text