Thermo Scientific Chemicals

Gadolinium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013207.22
100 g, Each
Thermo Scientific Chemicals

Gadolinium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013207.22
100 g, Each
Quantity
Catalog number: 013207.22
also known as 013207-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38245-36-2
IUPAC Name
digadolinium(3+) tricarbonate
Molecular Formula
C3Gd2O9
InChI Key
RQXZRSYWGRRGCD-UHFFFAOYSA-H
SMILES
[Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Gadolinium(III) carbonate is used for making optical glass and dopant for Gadolinium Yttrium Garnets which have microwave applications. It is also used for making phosphors for color TV tube and energy-saving lamps. It is used as intravenous MRI contrast agent to enhance images in medical magnetic resonance imaging and Magnetic Resonance Angiography (MRA) procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text