Thermo Scientific Chemicals

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013209.06
5 g, Each
Thermo Scientific Chemicals

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013209.06
5 g, Each
Quantity
Catalog number: 013209.06
also known as 013209-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
181266-82-0
IUPAC Name
europium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21EuO6
InChI Key
YMTKZYSHNNXTIJ-UHFFFAOYSA-N
SMILES
[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text