Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
5 g
Catalog number 013209.09
also known as 013209-09
Price (USD)/ Each
417.65
Online exclusive
464.00 
Save 46.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
417.65
Online exclusive
464.00 
Save 46.35 (10%)
Add to cart
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number013209.09
Price (USD)/ Each
417.65
Online exclusive
464.00 
Save 46.35 (10%)
-
Add to cart
Chemical Identifiers
CAS181266-82-0
IUPAC Nameeuropium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21EuO6
InChI KeyYMTKZYSHNNXTIJ-UHFFFAOYSA-N
SMILES[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only