Europium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Europium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 013210.14
also known as 013210-14
Price (USD)/ Each
505.65
Online exclusive
562.00 
Save 56.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
505.65
Online exclusive
562.00 
Save 56.35 (10%)
Add to cart
Europium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Catalog number013210.14
Price (USD)/ Each
505.65
Online exclusive
562.00 
Save 56.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10031-55-7
IUPAC Namedieuropium(3+) octahydrate trisulfate
Molecular FormulaEu2H16O20S3
InChI KeyPGDXJFDXKJLVTJ-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Eu+3].[Eu+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only