Thermo Scientific Chemicals

Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013214.09
10 g, Each
Thermo Scientific Chemicals

Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013214.09
10 g, Each
Quantity
Catalog number: 013214.09
also known as 013214-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14553-08-3
IUPAC Name
erbium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21ErO6
InChI Key
FGQSJRDKBCVFHH-UHFFFAOYSA-N
SMILES
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text