Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 013233.06
also known as 013233-06
Price (USD)/ Each
30.65
Online exclusive
33.60 
Save 2.95 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
33.60 
Save 2.95 (9%)
Add to cart
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number013233.06
Price (USD)/ Each
30.65
Online exclusive
33.60 
Save 2.95 (9%)
-
Add to cart
Chemical Identifiers
CAS12260-45-6
IUPAC Namecaesium(1+) hydroxide
Molecular FormulaCsHO
InChI KeyHUCVOHYBFXVBRW-UHFFFAOYSA-M
SMILES[OH-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only