Zinc sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Zinc sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc sulfide, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 013239.18
also known as 013239-18
Price (USD)/ Each
54.70
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
54.70
Add to cart
Zinc sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number013239.18
Price (USD)/ Each
54.70
-
Add to cart
Chemical Identifiers
CAS1314-98-3
IUPAC Namezinc(2+) sulfanediide
Molecular FormulaSZn
InChI KeyDRDVZXDWVBGGMH-UHFFFAOYSA-N
SMILES[S--].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Ziinc sulfide is used in luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only