Molybdenum pellet, 99.7% (metals basis), Thermo Scientific Chemicals
Molybdenum pellet, 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum pellet, 99.7% (metals basis), Thermo Scientific Chemicals

Approx. 12.5 x 18 x 25mm, HotPressed, density Approx. 8.0g/cm3
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number 013242.36
also known as 013242-36
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
Add to cart
Molybdenum pellet, 99.7% (metals basis), Thermo Scientific Chemicals
Catalog number013242.36
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only