Copper plate, 6.35mm (0.25in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Copper plate, 6.35mm (0.25in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper plate, 6.35mm (0.25in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx. 569g/10 x 10cm
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 x 20 cm
10 x 10 cm
Catalog number 013376.CU
also known as 013376-CU
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
20 x 20 cm
Request bulk or custom format
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
Add to cart
Copper plate, 6.35mm (0.25in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number013376.CU
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormPlate, 6.35mm (0.25in) thick.
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only