Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
20 x 100 cm
20 x 20 cm
Catalog number 013377.CV
also known as 013377-CV
Price (USD)/ Each
418.00
-
Add to cart
Quantity:
20 x 100 cm
Request bulk or custom format
Price (USD)/ Each
418.00
Add to cart
Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number013377.CV
Price (USD)/ Each
418.00
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay from Supplier's CofA≥99.9% (metals basis)
FormFoil, 1.27mm (0.05in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only