Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals
Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals

As a defoliant and epoxidation catalyst, as denaturing agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013395.AB
also known as 013395-AB
Price (USD)/ Each
171.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
202.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (USD)/ Each
171.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
202.00 
Save 30.35 (15%)
Add to cart
Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals
Catalog number013395.AB
Price (USD)/ Each
171.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
202.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1184-57-2
IUPAC Namemethylmercurylium hydroxide
Molecular FormulaCH4HgO
InChI KeyKRZWEBVPFGCYMY-UHFFFAOYSA-M
SMILES[OH-].C[Hg+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA1.0-1.05 M
Methyl mercury(II) hydroxide is used as defoliant and epoxidation catalyst, as denaturing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only