Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm, Thermo Scientific Chemicals
Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 013398.22
also known as 013398-22
Price (USD)/ Each
447.65
Online exclusive
497.00 
Save 49.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
447.65
Online exclusive
497.00 
Save 49.35 (10%)
Add to cart
Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm, Thermo Scientific Chemicals
Catalog number013398.22
Price (USD)/ Each
447.65
Online exclusive
497.00 
Save 49.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-35-8
IUPAC Nametrioxotungsten
Molecular FormulaO3W
InChI KeyZNOKGRXACCSDPY-UHFFFAOYSA-N
SMILESO=[W](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Mo0.002% max.
Impurity contentMo 100ppm
Tungsten(VI) oxide is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of tungsten(VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only