Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Diamond powder is generally used as an abrasive to grind and polish hard surfaces
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013402.03
also known as 013402-03
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
Add to cart
Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number013402.03
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7782-40-3
Specifications
Chemical Name or MaterialDiamond powder
Name NoteSynthetic
CAS7782-40-3
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only