Thermo Scientific Chemicals

Lithium sulfate, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 013404.30
250 g, Each
Thermo Scientific Chemicals

Lithium sulfate, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 013404.30
250 g, Each
Quantity
Catalog number: 013404.30
also known as 013404-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-48-7
IUPAC Name
dilithium(1+) sulfate
Molecular Formula
Li2O4S
InChI Key
INHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]S([O-])(=O)=O
Appearance (Color)
White
Form
Powder
Loss on Drying
1.0% max
Total Metal Impurities
0.3% max.

Description

Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text