Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 013405.30
250 g, Each
Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 013405.30
250 g, Each
Quantity
Catalog number: 013405.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-69-4
IUPAC Name
lithium(1+) nitrate
Molecular Formula
LiNO3
InChI Key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES
[Li+].[O-][N+]([O-])=O
Appearance (Color)
White
Form
Crystalline powder or powder or crystals
Assay from Suppliers CofA
≥98.5%

Description

Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text