Lithium titanium oxide, Thermo Scientific Chemicals
Lithium titanium oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium titanium oxide, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 013411.18
also known as 013411-18
Price (USD)/ Each
76.65
Online exclusive
85.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
85.10 
Save 8.45 (10%)
Add to cart
Lithium titanium oxide, Thermo Scientific Chemicals
Catalog number013411.18
Price (USD)/ Each
76.65
Online exclusive
85.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS12031-82-2
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max.
Lithium titanium oxide is used in battery, porcelain enamels and ceramic insulating bodies. It is frequently utilized as a flux due to its good stability. It is used as a cathode in layer one of a double layer cathode for molten carbonate fuel cells.