Lithium titanium oxide, Thermo Scientific Chemicals
Lithium titanium oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium titanium oxide, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 013411.30
also known as 013411-30
Price (USD)/ Each
303.65
Online exclusive
337.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
303.65
Online exclusive
337.00 
Save 33.35 (10%)
Add to cart
Lithium titanium oxide, Thermo Scientific Chemicals
Catalog number013411.30
Price (USD)/ Each
303.65
Online exclusive
337.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12031-82-2
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max.
Lithium titanium oxide is used in battery, porcelain enamels and ceramic insulating bodies. It is frequently utilized as a flux due to its good stability. It is used as a cathode in layer one of a double layer cathode for molten carbonate fuel cells.