Lithium aluminum oxide, Thermo Scientific Chemicals
Lithium aluminum oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium aluminum oxide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 013412.30
also known as 013412-30
Price (USD)/ Each
490.65
Online exclusive
545.00 
Save 54.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
490.65
Online exclusive
545.00 
Save 54.35 (10%)
Add to cart
Lithium aluminum oxide, Thermo Scientific Chemicals
Catalog number013412.30
Price (USD)/ Each
490.65
Online exclusive
545.00 
Save 54.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12003-67-7
IUPAC Name[(oxidoalumanylidene)oxo]lithiumylium
Molecular FormulaAlLiO2
InChI KeyYQNQTEBHHUSESQ-UHFFFAOYSA-N
SMILES[Li+].[O-][Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white
Formcrystalline solid
It is used as an inert electrolyte support material in molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only