Thermo Scientific Chemicals

Lithium acetate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 013417.A1
1 kg, Each
Thermo Scientific Chemicals

Lithium acetate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 013417.A1
1 kg, Each
Quantity
Catalog number: 013417.A1
also known as 013417-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6108-17-4
IUPAC Name
lithium(1+) acetate dihydrate
Molecular Formula
C2H7LiO4
InChI Key
IAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES
[Li+].O.O.CC([O-])=O
Insoluble material
0.005% max
pH
of a 5% solution 5.0-8.0
Trace Metals
(as Pb) 0.001% max
Impurity content
Cl 0.002% max
Impurity content
Fe 0.001% max

Description

Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text