Thermo Scientific Chemicals

Lithium carbonate, 99%, Thermo Scientific Chemicals

Catalog number: 013418.36
500 g, Each
Thermo Scientific Chemicals

Lithium carbonate, 99%, Thermo Scientific Chemicals

Catalog number: 013418.36
500 g, Each
Quantity
Catalog number: 013418.36
also known as 013418-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-13-2
IUPAC Name
dilithium(1+) carbonate
Molecular Formula
CLi2O3
InChI Key
XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]C([O-])=O
Assay (unspecified)
98.6% min

Description

Lithium carbonate-based glasses are used in overware. Used in the manufacture of lithium-ion battery cathodes. Pharmaceutical composition of lithium carbonate has been shown to exhibit antimanic effects specifically to treat mania associated with bipolar disorder. Used in both low-fire and high-fire ceramic glaze, tile adhesives, and in the processing of metal oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text