Thermo Scientific Chemicals

Lithium peroxide, 95%, Thermo Scientific Chemicals

Catalog number: 013421.22
100 g, Each
Thermo Scientific Chemicals

Lithium peroxide, 95%, Thermo Scientific Chemicals

Catalog number: 013421.22
100 g, Each
Quantity
Catalog number: 013421.22
also known as 013421-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12031-80-0
IUPAC Name
dilithium(1+) dioxidanediide
Molecular Formula
Li2O2
InChI Key
HPGPEWYJWRWDTP-UHFFFAOYSA-N
SMILES
[Li+].[Li+].[O-][O-]
Assay (unspecified)
90% min

Description

Lithium peroxide is used in air purifiers. It is also used to produce high-purity oxygen. It acts as a catalyst for polymerization of styrene to polystyrene. Further, it serves as a curing agent for special polymers and a source of oxygen in sealed spaces such as submarines and in breathing apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text