Thermo Scientific Chemicals

Lithium oxalate, 99+%, Thermo Scientific Chemicals

Catalog number: 013426.30
250 g, Each
Thermo Scientific Chemicals

Lithium oxalate, 99+%, Thermo Scientific Chemicals

Catalog number: 013426.30
250 g, Each
Quantity
Catalog number: 013426.30
also known as 013426-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
553-91-3
IUPAC Name
dilithium(1+) oxalate
Molecular Formula
C2Li2O4
InChI Key
YNQRWVCLAIUHHI-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]C(=O)C([O-])=O
Form
Powder
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥99.0%

Description

Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text