Thermo Scientific Chemicals

Lithium molybdenum oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 013427.36
500 g, Each
Thermo Scientific Chemicals

Lithium molybdenum oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 013427.36
500 g, Each
Quantity
Catalog number: 013427.36
also known as 013427-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13568-40-6
IUPAC Name
dilithium(1+) dioxomolybdenumbis(olate)
Molecular Formula
Li2MoO4
InChI Key
NMHMDUCCVHOJQI-UHFFFAOYSA-N
SMILES
[Li+].[Li+].[O-][Mo]([O-])(=O)=O
Impurity content
Ammonia: ≤0.0005%
Impurity content
Calcium: Report
Impurity content
Iron: ≤5 ppm
Impurity content
Sulfate: ≤0.01%
Water Content
Moisture: Report

Description

Lithium molybdenum oxide is mainly used as an inhibitor in some types of industrial air conditioning. It is used in comparative property studies of battery systems. It has been used as a petroleum cracking catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text