Thermo Scientific Chemicals

Sodium pyrophosphate, anhydrous, tech., Thermo Scientific Chemicals

Catalog number: 013439.30
250 g, Each
Thermo Scientific Chemicals

Sodium pyrophosphate, anhydrous, tech., Thermo Scientific Chemicals

Catalog number: 013439.30
250 g, Each
Quantity
Catalog number: 013439.30
also known as 013439-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7722-88-5
IUPAC Name
tetrasodium (phosphonatooxy)phosphonate
Molecular Formula
Na4O7P2
InChI Key
FQENQNTWSFEDLI-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
Assay (unspecified)
95% min

Description

Water softener, metal cleaner, soap and synthetic detergent builder, general sequestering agent, in electrodeposition of metals.Sodium pyrophosphate is used in water softener, detergent, emulsifying agent, metal cleaner and nutritional supplement. It acts as a buffering agent, thickening agent and dispersing agent. It also acts as a tartar control agent in toothpaste and dental floss. In addition, it is used as a chelating agent in antimicrobial studies. It is also used as a food additive in common foods such as chicken nuggets, crab meat and canned tuna.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Water softener, metal cleaner, soap and synthetic detergent builder, general sequestering agent, in electrodeposition of metals
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text