Ammonium dichromate, ACS, 99.5% min, Thermo Scientific Chemicals
Ammonium dichromate, ACS, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium dichromate, ACS, 99.5% min, Thermo Scientific Chemicals

Catalysts, source of pure nitrogen, in lithography
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013444.36
also known as 013444-36
Price (USD)/ Each
84.65
Online exclusive
93.80 
Save 9.15 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
93.80 
Save 9.15 (10%)
Add to cart
Ammonium dichromate, ACS, 99.5% min, Thermo Scientific Chemicals
Catalog number013444.36
Price (USD)/ Each
84.65
Online exclusive
93.80 
Save 9.15 (10%)
-
Add to cart
Chemical Identifiers
CAS7789-09-5
IUPAC Namediammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2H8N2O7
InChI KeyJOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% (NH₄)₂Cr₂O₇ min
Impurity contentCa 0.002%
Impurity contentCl 0.005%
Impurity contentFe 0.002%
Impurity contentNa 0.005%
View more
Catalysts, source of pure nitrogen, in lithography, manufacturing of pigmentsAmmonium dichromate is used in photo engraving, dye mordant, pigments, pickling, photography, process engraving, lithography, pyrotechnics, glazes for porcelain and china wares. It acts as a catalyst and a source of nitrogen in the laboratory and as the photoactive site. It is also used in the preparation of alizarin and chrome alum. Further, it is used in leather and oil industry for tanning and purification respectively. It is involved in the production of the phosphor raster television screens and other devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only