Thermo Scientific Chemicals

Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%, Thermo Scientific Chemicals

Catalog number: 013448.A3
2 kg, Each
Thermo Scientific Chemicals

Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%, Thermo Scientific Chemicals

Catalog number: 013448.A3
2 kg, Each
Quantity
Catalog number: 013448.A3
also known as 013448-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-85-9
IUPAC Name
λ²-iron(2+) diammonium hexahydrate disulfate
Molecular Formula
FeH20N2O14S2
InChI Key
MQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Impurity content
Cu 0.003%
Impurity content
Ferric ion (Fe⁺³) 0.01%
Impurity content
Na 0.02%
Assay (unspecified)
98.5-101.5% H₈FeN₂O₈S₂.6H₂O
Impurity content
Ca 0.005%

Description

Polymerization catalyst, photographyAmmonium iron(II) sulfate hexahydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text