Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals
Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
5 x 2 kg
Catalog number 013449.36
also known as 013449-36
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
Add to cart
Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals
Catalog number013449.36
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
-
Add to cart
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.005%
Impurity contentCa 0.005%
Assay (unspecified)99% C₂H₃KO₂ min
Heavy Metals(ICP-OES) 5ppm
Impurity contentCl 0.003%
View more
Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only