Thermo Scientific Chemicals

Potassium dichromate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 013450.22
100 g, Each
Thermo Scientific Chemicals

Potassium dichromate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 013450.22
100 g, Each
Quantity
Catalog number: 013450.22
also known as 013450-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7778-50-9
IUPAC Name
dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular Formula
Cr2K2O7
InChI Key
KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES
[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
Impurity content
Cl 0.001%
Impurity content
Fe 0.001%
Impurity content
Na 0.02%
Impurity content
SO₄ 0.005%
Insoluble material
0.005%

Description

As an oxidizing agent, in dyeing, painting, decorating porcelain, lithography, in electric batteries, electroplating Potassium dichromate is most often used as an oxidizing agent in laboratories and industries. It is used as a precursor to prepare potassium chrome alum which finds application in leather finishing and tanning. It is utilized in the preparation of chromic acid for cleaning glassware and etching materials It is an active ingredient in cement and is used for retards setting and improves its density and texture. Furthermore, it is used in photography and in photographic screen printing. It is also used in wet test analysis as an analytical lab reagent and in ethanol determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Insoluble matter 0.005%, Loss on drying 0.05%, Cl 0.001%, SO4 0.005%, Ca 0.003%, Fe 0.001%, Na 0.02%
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text