Thermo Scientific Chemicals

Potassium hydroxide, ACS, 85% min, K{2}CO{3} 2.0% max, Thermo Scientific Chemicals

Catalog number: 013451.18
50 g, Each
Thermo Scientific Chemicals

Potassium hydroxide, ACS, 85% min, K{2}CO{3} 2.0% max, Thermo Scientific Chemicals

Catalog number: 013451.18
50 g, Each
Quantity
Catalog number: 013451.18
also known as 013451-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-58-3
IUPAC Name
potassium hydroxide
Molecular Formula
HKO
InChI Key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES
[OH-].[K+]
Assay (unspecified)
85% KOH min
Heavy Metals
(ICP-OES) 0.001%
Impurity content
2.0% K₂CO₃ max
Impurity content
Ca 0.005%
Impurity content
Ni 0.001%

Description

In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagentPotassium hydroxide acts as a precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further, it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text