Thermo Scientific Chemicals

Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals

Catalog number: 013452.22
100 g, Each
Thermo Scientific Chemicals

Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals

Catalog number: 013452.22
100 g, Each
Quantity
Catalog number: 013452.22
also known as 013452-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6487-48-5
IUPAC Name
dipotassium hydrate oxalate
Molecular Formula
C2H2K2O5
InChI Key
QCPTVXCMROGZOL-UHFFFAOYSA-L
SMILES
O.[K+].[K+].[O-]C(=O)C([O-])=O
Comment
Neutrality P.T.
Heavy Metals
(as Pb) 0.002%
Impurity content
Cl 0.002%
Impurity content
Fe 0.001%
Impurity content
Na 0.02%

Description

Potassium oxalate monohydrate is used as a chelating agent; a purifying agent in pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, a rust remover for metal treatment, a bleaching agent in textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as a reducing agent in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text