Thermo Scientific Chemicals

Sodium hydroxide (low chloride), ACS, 97.0% min, Thermo Scientific Chemicals

Catalog number: 013455.F6
2 x 12.5 kg, Each
Thermo Scientific Chemicals

Sodium hydroxide (low chloride), ACS, 97.0% min, Thermo Scientific Chemicals

Catalog number: 013455.F6
2 x 12.5 kg, Each
Quantity
Catalog number: 013455.F6
also known as 013455-F6
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Impurity content
Ca 0.005%
Impurity content
Cl 0.005%
Impurity content
Fe 0.001%
Impurity content
Hg 0.1ppm
Impurity content
K 0.02%

Description

In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal saltsSodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text