Thermo Scientific Chemicals
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number: 013476.RR
10 x 10 cm, Each
Thermo Scientific Chemicals
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number: 013476.RR
10 x 10 cm, Each
Quantity
Catalog number: 013476.RR
also known as 013476-RR
Price (USD)
Quantity
-
Have Questions?
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.