Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 60 cm
10 x 10 cm
20 x 30 cm
Catalog number 013476.RU
also known as 013476-RU
Price (USD)/ Each
276.00
-
Add to cart
Quantity:
30 x 60 cm
Request bulk or custom format
Price (USD)/ Each
276.00
Add to cart
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number013476.RU
Price (USD)/ Each
276.00
-
Add to cart
Specifications
Chemical Name or MaterialStainless Steel gauze
Name NoteOpen area: 36%; Width of opening: 0.003 in.
Health Hazard 1H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
Recommended StorageAmbient temperatures
TSCAYes
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only