Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005in) dia wire, Type 304, Thermo Scientific Chemicals
Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005in) dia wire, Type 304, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005in) dia wire, Type 304, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 x 10 cm
20 x 30 cm
30 x 60 cm
Catalog number 013477.RR
also known as 013477-RR
Price (USD)/ Each
57.90
-
Add to cart
Quantity:
10 x 10 cm
Request bulk or custom format
Price (USD)/ Each
57.90
Add to cart
Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number013477.RR
Price (USD)/ Each
57.90
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialStainless Steel gauze, Type 304
Name NoteWoven from 0.127mm (0.005 in.) dia. wire
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 70:19:11 wt%
TSCAYes
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.