Thermo Scientific Chemicals
Stainless Steel gauze, 40 mesh woven from 0.25mm (0.01in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number: 013478.CH
20 x 30 cm, Each
Thermo Scientific Chemicals
Stainless Steel gauze, 40 mesh woven from 0.25mm (0.01in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number: 013478.CH
20 x 30 cm, Each
Quantity
Catalog number: 013478.CH
also known as 013478-CH
Price (USD)
Quantity
-
Have Questions?
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.