Thermo Scientific Chemicals

Rubidium hydroxide hydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013481.14
25 g, Each
Thermo Scientific Chemicals

Rubidium hydroxide hydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013481.14
25 g, Each
Quantity
Catalog number: 013481.14
also known as 013481-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12026-05-0
IUPAC Name
rubidium(1+) hydroxide
Molecular Formula
HORb
InChI Key
CPRMKOQKXYSDML-UHFFFAOYSA-M
SMILES
[OH-].[Rb+]
Total Metal Impurities
1% max.

Description

Oxidative chlorination catalyst.Rubidium hydroxide hydrate is used in fireworks in place of pure rubidium to get violet color, thereby it prevents the waste of expensive element rubidium. It acts as an intermediate in the synthesis of other rubidium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text