Thermo Scientific Chemicals

Mercury(II) sulfide, red, Thermo Scientific Chemicals

Catalog number: 013482.36
500 g, Each
Thermo Scientific Chemicals

Mercury(II) sulfide, red, Thermo Scientific Chemicals

Catalog number: 013482.36
500 g, Each
Quantity
Catalog number: 013482.36
also known as 013482-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1344-48-5
IUPAC Name
mercury(2+) sulfanediide
Molecular Formula
HgS
InChI Key
LGZXYFMMLRYXLK-UHFFFAOYSA-N
SMILES
[S--].[Hg++]
Assay (unspecified)
99% min

Description

Mercury(II) sulfide, red is used chiefly as a pigment in paints, rubber and plastics. It is also used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text