Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals
Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 013493.14
also known as 013493-14
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
Add to cart
Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals
Catalog number013493.14
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7488-54-2
IUPAC Namesulfuric acid rubidium
Molecular FormulaH2O4RbS
InChI KeyXNNUAZGCOYTNQE-UHFFFAOYSA-N
SMILES[Rb].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only