Thermo Scientific Chemicals

Rubidium iodide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 013497.09
10 g, Each
Thermo Scientific Chemicals

Rubidium iodide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 013497.09
10 g, Each
Quantity
Catalog number: 013497.09
also known as 013497-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-29-6
IUPAC Name
rubidium(1+) iodide
Molecular Formula
IRb
InChI Key
WFUBYPSJBBQSOU-UHFFFAOYSA-M
SMILES
[Rb+].[I-]
Total Metal Impurities
0.2% max.

Description

Rubidium iodide is used for making metal rubidium and other rubidium salts. It is also used in the production of special glass, miniature high energy battery and crystal scintillation counter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text