Thermo Scientific Chemicals

Lithium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 013600.06
5 g, Each
Thermo Scientific Chemicals

Lithium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 013600.06
5 g, Each
Quantity
Catalog number: 013600.06
also known as 013600-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-51-2
IUPAC Name
lithium(1+) iodide
Molecular Formula
ILi
InChI Key
HSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES
[Li+].[I-]
Appearance (Color)
White
Form
Beads
Particle Size
-10 Mesh
Residual water
< 100 ppm
Total Metal Impurities
0.001% max.

Description

It is used as an absorbent in refrigeration process, as a catalyst, and in photography. It is also useful for organic synthesis, pessat iodination, cleavage of esters, ethers, epoxides, decarboethoxylation, carbonylation, and aldol condensation. It is used for manufacturing medicines and mineral waters. It is used in electochemicstry and redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text