Thermo Scientific Chemicals

Tin(IV) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 013617.14
25 g, Each
Thermo Scientific Chemicals

Tin(IV) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 013617.14
25 g, Each
Quantity
Catalog number: 013617.14
also known as 013617-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-47-8
IUPAC Name
tin(IV) iodide
Molecular Formula
H4I4Sn
InChI Key
ZBCVLXKKLMNLGV-UHFFFAOYSA-N
SMILES
[Sn].I.I.I.I
Appearance (Color)
Orange to Red
Form
Powder
Residual water
< 100 ppm
Total Metal Impurities
0.002% max.

Description

Tin(IV) iodide is used in the preparation of tin(IV) oxide and tin(II) oxide. It reacts with hydroiodic acid to get rare tin hexaiodide. It forms tetraiodobis triphenylphosphine tin complexes by reacting with triphenylphosphine, which finds application in toothpaste formulation. Further, it acts as analytical reagents and also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text