Thermo Scientific Chemicals

Iron(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013619.09
10 g, Each
Thermo Scientific Chemicals

Iron(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013619.09
10 g, Each
Quantity
Catalog number: 013619.09
also known as 013619-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-86-0
IUPAC Name
λ²-iron(2+) diiodide
Molecular Formula
FeI2
InChI Key
BQZGVMWPHXIKEQ-UHFFFAOYSA-L
SMILES
[Fe++].[I-].[I-]
Residual water
< 100 ppm
Total Metal Impurities
0.01% max.
Particle Size
-10 Mesh
Appearance (Color)
Red-brown
Form
Beads

Description

Iron(II) iodide is used as a catalyst in organic reactions. It is also used as a compound in semiconductors and other high purity applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text