Thermo Scientific Chemicals

Cerium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013641.03
1 g, Each
Thermo Scientific Chemicals

Cerium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013641.03
1 g, Each
Quantity
Catalog number: 013641.03
also known as 013641-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-87-6
IUPAC Name
cerium(3+) triiodide
Molecular Formula
CeI3
InChI Key
ZEDZJUDTPVFRNB-UHFFFAOYSA-K
SMILES
[I-].[I-].[I-].[Ce+3]
Appearance (Color)
Yellow to yellow-green to brown
Form
Powder
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.1% max.

Description

Cerium(III) iodide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text