Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number 013650.14
also known as 013650-14
Price (USD)/ Each
67.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
67.20
Add to cart
Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Catalog number013650.14
Price (USD)/ Each
67.20
-
Add to cart
Chemical Identifiers
CAS13709-49-4
IUPAC Nameyttrium(3+) trifluoride
Molecular FormulaF3Y
InChI KeyRBORBHYCVONNJH-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Suppliers CofA≥99+% (based on elemental Yttrium)
Elemental AnalysisYttrium (Y): ≥59.72%
Yttrium(III) fluoride is used in the production of yttrium metal, glasses and ceramics. It is also used in the preparation of low-index film layers, which exhibit good transparency in the UV through infrared regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only