Thermo Scientific Chemicals

Thulium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013652.14
25 g, Each
Thermo Scientific Chemicals

Thulium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013652.14
25 g, Each
Quantity
Catalog number: 013652.14
also known as 013652-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13760-79-7
IUPAC Name
thulium(3+) trifluoride
Molecular Formula
F3Tm
InChI Key
AATUHDXSJTXIHB-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Tm+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Thulium(III) fluoride, anhydrous is used for glass doping and to enhance mid-IR emission in oxyfluoride glass ceramics. It is also used to improve ultraviolet and visible upconversion fluorescence in fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text