Thermo Scientific Chemicals

Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013657.06
5 g, Each
Thermo Scientific Chemicals

Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013657.06
5 g, Each
Quantity
Catalog number: 013657.06
also known as 013657-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13708-63-9
IUPAC Name
terbium(3+) trifluoride
Molecular Formula
F3Tb
InChI Key
LKNRQYTYDPPUOX-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Tb+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text