Thermo Scientific Chemicals

Lanthanum(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013663.18
50 g, Each
Thermo Scientific Chemicals

Lanthanum(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 013663.18
50 g, Each
Quantity
Catalog number: 013663.18
also known as 013663-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13709-38-1
IUPAC Name
lanthanum(3+) trifluoride
Molecular Formula
F3La
InChI Key
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[La+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lanthanum fluoride, anhydrous is employed extensively in the preparation and study of fluoride glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text